Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Pro EGF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5256SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5256SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5256SP Supplier R&D Systems Supplier No. MAB5256SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

EGF Monoclonal specifically detects EGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen EGF
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 463004
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01132
Gene Alias beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone
Gene Symbols EGF
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EGF Trp29-Arg1029 Accession # P01132
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1950
Test Specificity Detects mouse Pro EGF in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Amphiregulin, rmEpigen, rmEpiregulin, rmEGF, rmMFG-E8, recombinant human (rh) EGF, rhHB-EGF, rhHRG alpha, rhHRG beta, or recombinant rat EGF is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.