Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Pro-MMP-7 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB29671
This item is not returnable.
View return policy
Description
MMP-7 Monoclonal specifically detects MMP-7 in Mouse samples. It is validated for Western Blot.Specifications
MMP-7 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
AAA99983 | |
MMP7 | |
Mouse myeloma cell line NS0-derived recombinant mouse MMP-7 Leu18-Leu264 Accession # AAA99983 | |
100 μg | |
Primary | |
Detects the pro form of mouse MMP-7 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse MMP-2, -3, -8, -9, -12, or -24 is observed. | |
Mouse | |
Purified |
Western Blot | |
377313 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.24, EC 3.4.24.23, matrilysin, Matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4316 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction