Learn More
Novus Biologicals™ Mouse Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Description
Specifications
Specifications
| Assay Range | 93.75 to 6000 pg/mL |
| Assay Sensitivity | 56.25 pg/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Biological Fluids, Plasma, Serum |
| Specificity | The Mouse Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric) recognizes Mouse P1NP in samples. No significant cross-reactivity or interference between Mouse P1NP and analogues was observed. |
| Gene ID (Entrez) | 1277 |
| Host Species | Mouse |
| Quantity | 1 Kit |
| Research Discipline | Cellular Markers, Extracellular Matrix, Metastasis |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.