Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Progranulin/PGRN Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB25572100
This item is not returnable.
View return policy
Description
Progranulin/PGRN Monoclonal specifically detects Progranulin/PGRN in Mouse samples. It is validated for ELISA.Specifications
| Progranulin/PGRN | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Acrogranin, CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN, Proepithelin | |
| Mouse myeloma cell line NS0-derived recombinant mouse Progranulin/PGRN Thr18-Leu589 Accession # P28798 | |
| 100 μg | |
| 2896 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 333729R | |
| ELISA | |
| P28798 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects mouse Progranulin/PGRN in direct ELISAs. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction