Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Prostasin/Prss8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2968SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2968SP 25 μg
AF2968 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2968SP Supplier R&D Systems Supplier No. AF2968SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Prostasin/Prss8 Polyclonal specifically detects Prostasin/Prss8 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Prostasin/Prss8
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99L44
Gene Alias CAP1, channel-activating protease 1, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.120, prostasin, protease, serine, 8, Prss8, Serine protease 8
Gene Symbols PRSS8
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Prostasin/Prss8 Ala30-Gln289 Accession # Q99L44
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5652
Test Specificity Detects mouse Prostasin/Prss8 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity is observed with recombinant human Prostasin.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.