Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Protein Disulfide Isomerase/P4HB ELISA Kit (Colorimetric)
SDP

Catalog No. NB016782 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016782 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016782 Supplier Novus Biologicals™ Supplier No. NBP281223
Add to Cart
Add to Cart

This kit recognizes Mouse Protein Disulfide Isomerase/P4HB in samples.

No significant cross-reactivity or interference between Mouse Protein Disulfide Isomerase/P4HB and analogues was observed.

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.09 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 5034
Gene Symbol P4HB
Host Species Mouse
Quantity 1 Kit
Research Discipline Cellular Markers, ER Markers, Fibroblast Cell Markers, Hypoxia
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Protein Disulfide Isomerase/P4HB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.