Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Protocadherin-12 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7926SP
This item is not returnable.
View return policy
Description
Protocadherin-12 Monoclonal specifically detects Protocadherin-12 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Protocadherin-12 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
O55134 | |
PCDH12 | |
Mouse myeloma cell line NS0-derived recombinant mouse Protocadherin-12 Met1-Ala716 Accession # O55134 | |
25 μg | |
Primary | |
Detects mouse Protocadherin-12 in ELISA. | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
868909 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PCDH12, protocadherin 12, Protocadherin12, Vascular cadherin-2, vascular endothelial cadherin 2, Vascular endothelial cadherin-2, VE-cad-2, VECAD2, VE-cadherin 2, VE-cadherin-2, VE-cadherin-2protocadherin-12 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
51294 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction