Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Protogenin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4919SP
This item is not returnable.
View return policy
Description
Protogenin Polyclonal specifically detects Protogenin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Protogenin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ12795, FLJ13221, FLJ25756, IGDCC5, immunoglobulin superfamily, DCC subclass, member 5, Protein Shen-Dan, protogenin, protogenin homolog (Gallus gallus), PRTG, RPTG, Shen-Dan | |
Sheep | |
Affinity Purified | |
RUO | |
283659 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q2EY15 | |
PRTG | |
Mouse myeloma cell line NS0-derived recombinant mouse PRTG Val24-Thr943 Accession # Q2EY15 | |
25 μg | |
Primary | |
Detects mouse and human PRTG in Western blots. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction