Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PU.1/Spi-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7124
This item is not returnable.
View return policy
Description
PU.1/Spi-1 Polyclonal specifically detects PU.1/Spi-1 in Mouse samples. It is validated for Western Blot.Specifications
PU.1/Spi-1 | |
Polyclonal | |
Unconjugated | |
P17433 | |
Spi1 | |
E. coli-derived recombinant mouse PU.1/Spi-1 Met1-Lys169 Accession # P17433 | |
100 μg | |
Primary | |
Detects mouse PU.1/Spi-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human Spi-1 is observed, and less than 1% cross-reactivity with recombinant mouse Spi-B is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
hematopoietic transcription factor PU.1, OF, PU.1, SFPI1, SFPI131 kDa transforming protein, SPI1, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1 | |
Sheep | |
Affinity Purified | |
RUO | |
6688 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction