Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Pygopus-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $468.00
Specifications
Antigen | Pygopus-1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Pygopus-1 Polyclonal specifically detects Pygopus-1 in Mouse samples. It is validated for Western Blot.Specifications
Pygopus-1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
DKFZp547G0910, Pygo1, pygopus 1, pygopus homolog 1, pygopus homolog 1 (Drosophila), Pygopus1, pygopus-like protein 1 | |
PYGO1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9D0P5 | |
26108 | |
E. coli-derived recombinant mouse Pygopus-1 Ser2-Ala417 Accession # Q9D0P5 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Pygopus-1 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title