Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Rae-1 epsilon Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1135N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1135N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1135N100 Supplier R&D Systems Supplier No. FAB1135N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Rae-1 epsilon Monoclonal specifically detects Rae-1 epsilon in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Rae-1 epsilon
Applications Flow Cytometry
Classification Monoclonal
Clone 205001
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Symbols Raet1e
Host Species Rat
Immunogen BaF3 mouse pro-B cell line transfected with mouse Rae-1 epsilon
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects mouse Rae-1 epsilon in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Rae-1 alpha , beta , gamma , or delta is observed. By flow cytometry, no cross-reactivity with mouse Rae-1 alpha or mouse Rae-1 gamma .
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.