Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Rae-1 epsilon Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
$200.50 - $2020.00
Specifications
Antigen | Rae-1 epsilon |
---|---|
Clone | 205001 |
Concentration | LYOPH |
Dilution | Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready |
Applications | Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1135SP
![]() |
R&D Systems
MAB1135SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1135100
![]() |
R&D Systems
MAB1135100 |
100 μg |
Each for $549.50
|
|
|||||
MAB1135500
![]() |
R&D Systems
MAB1135500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
Rae-1 epsilon Monoclonal specifically detects Rae-1 epsilon in Mouse samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
Rae-1 epsilon | |
LYOPH | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Raet1e | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
205001 | |
Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
Monoclonal | |
Rat | |
Mouse | |
379043 | |
BaF3 mouse pro-B cell line transfected with mouse Rae-1 epsilon | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Rae-1 epsilon in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Rae-1 alpha , beta , gamma , or delta is observed. By flow cytometry, no cross-reactivity with mouse Rae-1 alpha or mouse Rae-1 gamma . |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title