Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse RAGE Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB11794
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11794 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB11794 Supplier R&D Systems Supplier No. MAB11794
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

AGER Monoclonal specifically detects AGER in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen RAGE
Applications Immunohistochemistry
Classification Monoclonal
Clone 697005
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q62151
Gene Alias advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1
Gene Symbols Ager
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse RAGE Gly23-Leu342 Accession # Q62151
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5891
Test Specificity Detects mouse RAGE in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant rat RAGE, 20% - 50% cross-reactivity with recombinant human RAGE, and no cross-reactivity with recombinant canine RAGE is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.