Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse RAMP2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB6500U100UG
This item is not returnable.
View return policy
Description
RAMP2 Monoclonal specifically detects RAMP2 in Mouse samples. It is validated for Flow Cytometry.Specifications
RAMP2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9WUP0 | |
RAMP2 | |
E. coli-derived recombinant mouse RAMP2 Ser45-Val159 Accession # Q9WUP0 | |
RUO | |
10266 | |
Mouse | |
IgG2a |
Flow Cytometry | |
773816 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
calcitonin receptor-like receptor activity modifying protein 2, Calcitonin-receptor-like receptor activity-modifying protein 2, CRLR activity-modifying protein 2, receptor (calcitonin) activity modifying protein 2, receptor (G protein-coupled) activity modifying protein 2, receptor activity modifying protein 2, receptor activity-modifying protein 2, receptor-activity-modifying protein 2 | |
Rat | |
100 μg | |
Primary | |
Detects mouse RAMP2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human RAMP2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction