Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse RAMP2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB6500SP
This item is not returnable.
View return policy
Description
RAMP2 Monoclonal specifically detects RAMP2 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
RAMP2 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
Q9WUP0 | |
RAMP2 | |
E. coli-derived recombinant mouse RAMP2 Ser45-Val159 Accession # Q9WUP0 | |
25 μg | |
Primary | |
Detects mouse RAMP2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human RAMP2 is observed. | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
773816 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
calcitonin receptor-like receptor activity modifying protein 2, Calcitonin-receptor-like receptor activity-modifying protein 2, CRLR activity-modifying protein 2, receptor (calcitonin) activity modifying protein 2, receptor (G protein-coupled) activity modifying protein 2, receptor activity modifying protein 2, receptor activity-modifying protein 2, receptor-activity-modifying protein 2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10266 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction