Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse RANK/TNFRSF11A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB692
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB692 500 μg
MAB692SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB692 Supplier R&D Systems Supplier No. MAB692
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

RANK/TNFRSF11A Monoclonal specifically detects RANK/TNFRSF11A in Mouse samples. It is validated for Western Blot.

Specifications

Antigen RANK/TNFRSF11A
Applications Western Blot
Classification Monoclonal
Clone 100024
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O35305
Gene Alias CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFR, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TNFRSF11A, TRANCE R, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Gene Symbols TNFRSF11A
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse RANK/TNFRSF11A Leu29-Pro213 (Leu29Met) Accession # O35305
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8792
Test Specificity Detects mouse RANK/TNFRSF11A in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) 4-1BB, rmBAFF R, rmCD27, rmCD30, rmCD40, rmDR3, rmDR6, rmEDAR, rmFAS, rmGITR, recombinant human (rh) HVEM, rmLT beta R, rmNGF R, rmOPG, rmOX40, rhRELT, rmTROY, rmTNF RI, or rmTNF RII is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.