Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse RARRES1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4657
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4657 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4657 Supplier R&D Systems Supplier No. AF4657
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Produced in goats immunized with purified, E. coli-derived, recombinant mouse RARRES1 (rmRARRES1; aa 42 - 283; Accession # XP_001475843). Mouse RARRES1 specific IgG was purified by mouse RARRES1 affinity chromatography.

RARRES1 Polyclonal specifically detects RARRES1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RARRES1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. F6ZIS7
Gene Alias RAR-responsive protein TIG1, retinoic acid receptor responder (tazarotene induced) 1, retinoic acid receptor responder protein 1, TIG1, TIG1Tazarotene-induced gene 1 protein
Gene Symbols RARRES1
Host Species Goat
Immunogen E. coli-derived recombinant mouse RARRES1 Pro42-Phe283 Accession # F6ZIS7
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5918
Test Specificity Detects mouse RARRES1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human RARRES1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.