Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat alpha 1B-Glycoprotein Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8537 Supplier R&D Systems Supplier No. AF8537
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

alpha 1B-Glycoprotein Polyclonal specifically detects alpha 1B-Glycoprotein in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Specifications

Antigen alpha 1B-Glycoprotein
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Simple Western 1:100 dilution, Immunocytochemistry 1:20-1:50 dilution
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. with Sodium Azide
Gene Accession No. Q19LI2
Gene Symbols A1bg
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse alpha 1B-Glycoprotein Leu21-Gly512 Accession # Q19LI2
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117586
Test Specificity Detects mouse and rat alpha 1B-Glycoprotein in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human alpha 1B-Glycoprotein observed.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.