Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat Apolipoprotein H/ApoH Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6575SP
This item is not returnable.
View return policy
Description
Apolipoprotein H/ApoH Polyclonal specifically detects Apolipoprotein H/ApoH in Mouse, Rat samples. It is validated for Western Blot.Specifications
Apolipoprotein H/ApoH | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, APOH, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG | |
Sheep | |
Affinity Purified | |
RUO | |
350 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.25 ug/mL | |
Q01339 | |
APOH | |
Mouse myeloma cell line NS0-derived recombinant mouse Apolipoprotein H/ApoH Gly20-Cys345 Accession # Q01339 | |
25 μg | |
Primary | |
Detects mouse and rat Apolipoprotein H/ApoH in Western blots and detects recombinant mouse Apolipoprotein H/ApoH in direct ELISAs. In direct ELISAs, approximately 40% cross-reactivity with recombinant human Apolipoprotein H/ApoH is observed. | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction