Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Apolipoprotein H/ApoH Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6575
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6575 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6575 Supplier R&D Systems Supplier No. AF6575
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Apolipoprotein H/ApoH Polyclonal specifically detects Apolipoprotein H/ApoH in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Apolipoprotein H/ApoH
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL
Gene Accession No. Q01339
Gene Alias Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, APOH, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG
Gene Symbols APOH
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Apolipoprotein H/ApoH Gly20-Cys345 Accession # Q01339
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 350
Test Specificity Detects mouse and rat Apolipoprotein H/ApoH in Western blots and detects recombinant mouse Apolipoprotein H/ApoH in direct ELISAs. In direct ELISAs, approximately 40% cross-reactivity with recombinant human Apolipoprotein H/ApoH is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.