Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat BMP-15/GDF-9B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5917
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5917 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5917 Supplier R&D Systems Supplier No. AF5917
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

BMP-15/GDF-9B Polyclonal specifically detects BMP-15/GDF-9B in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen BMP-15/GDF-9B
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q9Z0L4
Gene Alias BMP15, BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4
Gene Symbols BMP15
Host Species Sheep
Immunogen E. coli-derived recombinant mouse BMP-15/GDF-9B Gln268-Arg392 Accession # Q9Z0L4
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9210
Test Specificity Detects mouse and rat BMP-15/GDF-9B in Western blots and recombinant mouse in direct ELISAs. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) BMP-15 is observed and less than 1% cross-reactivity with rhBMP-3 and rhBMP-10 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.