Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat Butyrylcholinesterase/BCHE Antibody, R&D Systems™

Goat Polyclonal Antibody
$188.28 - $567.67
Specifications
| Antigen | Butyrylcholinesterase/BCHE |
|---|---|
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Butyrylcholinesterase/BCHE Polyclonal specifically detects Butyrylcholinesterase/BCHE in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Butyrylcholinesterase/BCHE | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Acylcholine acylhydrolase, BCHE, Butyrylcholine esterase, butyrylcholinesterase, CHE1, CHE1cholinesterase, Choline esterase II, cholinesterase 1, E1, EC 3.1.1.8, Pseudocholinesterase | |
| Bche | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse, Rat | |
| AAH99977 | |
| 590 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Butyrylcholinesterase/BCHE His28-Leu603 Accession # AAH99977 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse and rat Butyrylcholinesterase/BCHE in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title