Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Carboxylesterase 3/CES3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5985
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5985 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5985 Supplier R&D Systems Supplier No. AF5985
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Carboxylesterase 3/CES3/Esterase 31 Polyclonal specifically detects Carboxylesterase 3/CES3/Esterase 31 in Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Carboxylesterase 3/CES3/Esterase 31
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. Q8VCT4
Gene Alias carboxylesterase 3, carboxylesterase 3 (brain), CES3, EC 3.1.1, EC 3.1.1.1, ES31FLJ21736, esterase 31, Liver carboxylesterase 31 homolog
Gene Symbols CES3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Carboxylesterase 3/CES3 Tyr19-Glu561 Accession # Q8VCT4
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23491
Test Specificity Detects recombinant mouse Carboxylesterase 3/CES3 in direct ELISAs and Western blots. Detects intrinsic CES3 in mouse and rat tissues in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) CES1, rhCES2, recombinant mouse (rm) CES2, and rmCES5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.