Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat CCR8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5997
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5997 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5997 Supplier R&D Systems Supplier No. AF5997
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

CCR8 Polyclonal specifically detects CCR8 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen CCR8
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Gene Accession No. P56484
Gene Alias CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR8, CCR-8, CD198, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, ChemR1, CKRL1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCL1, GPRCY6, GPR-CY6, TER-1, TER1CC chemokine receptor CHEMR1
Gene Symbols CCR8
Host Species Sheep
Immunogen E. coli-derived recombinant mouse CCR8 Met1-Met33, Asn92-Lys105, Tyr170-Ile200, His262-Leu278 Accession # P56484
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1237
Test Specificity Detects mouse and rat CCR8 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant mouse (rm) CCR1, rmCCR4, and rmCCR5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.