Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat CD200 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems AF3355
This item is not returnable.
View return policy
Description
CD200/OX2 Polyclonal specifically detects CD200/OX2 in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
CD200/OX2 | |
Polyclonal | |
Unconjugated | |
O54901 | |
CD200 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD200 Gln31-Gly232 Accession # O54901 | |
100 μg | |
Primary | |
Detects mouse CD200 in direct ELISAs and mouse CD200 and rat CD200 Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human CD200 is observed. | |
Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
antigen identified by monoclonal MRC OX-2, CD200, CD200 antigenMOX1, CD200 molecule, MOX1, MOX2, MOX2MRC, MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
Goat | |
Affinity Purified | |
RUO | |
4345 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction