Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat CD31/PECAM-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 16 publications
$221.50 - $444.00
Specifications
Antigen | CD31/PECAM-1 |
---|---|
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Polyclonal |
Conjugate | Alexa Fluor 488 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB3628G025
![]() |
R&D Systems
FAB3628G025 |
25 Tests |
Each for $221.50
|
|
|||||
FAB3628G100
![]() |
R&D Systems
FAB3628G100 |
100 Tests |
Each for $444.00
|
|
|||||
Description
CD31/PECAM-1 Polyclonal specifically detects CD31/PECAM-1 in Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
CD31/PECAM-1 | |
Flow Cytometry | |
Alexa Fluor 488 | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
PECAM1 | |
IgG | |
Affinity Purified | |
Detects mouse CD31/PECAM-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CD31 and recombinant porcine CD31 is observed. Detects mouse CD31 and rat CD31 in flow cytometry. |
Flow Cytometry 5 uL/10^6 cells | |
Polyclonal | |
Goat | |
Mouse, Rat | |
Q08481 | |
5175 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD31/PECAM-1 Glu18-Lys590 Accession # Q08481 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title