Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat CD34 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF6518
This item is not returnable.
View return policy
Description
CD34 Polyclonal specifically detects CD34 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CD34 | |
Polyclonal | |
Unconjugated | |
Q64314 | |
CD34 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD34 Thr35-Thr287 Accession # Q64314 | |
100 μg | |
Primary | |
Detects mouse and rat CD34 in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant porcine CD34 is observed. | |
Mouse, Rat | |
IgG |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1 | |
Sheep | |
Affinity Purified | |
RUO | |
947 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction