Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat DARC APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6695A100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6695A100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6695A100 Supplier R&D Systems Supplier No. FAB6695A100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

DARC Polyclonal specifically detects DARC in Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen DARC
Applications Flow Cytometry
Classification Polyclonal
Conjugate APC
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied in a saline solution containing BSA and sodium azide
Gene Accession No. NP_034175
Gene Alias ACKR1, CCBP1, CD234, CD234 antigen, Dfy, Duffy Antigen, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy Antigen, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPD, GPDWBCQ1, GpFy, Plasmodium vivax receptor
Gene Symbols DARC
Host Species Sheep
Immunogen E. coli-derived recombinant mouse DARC, Met1-Pro61, Ala115-Cys127, Ser186-Lys205, Tyr264-Asn285, Accession No. NP_034175
Purification Method Antigen Affinity-purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2532.0
Test Specificity Detects mouse and rat DARC in Western blots.
Target Species Mouse, Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.