Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat DLL1 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF3970
This item is not returnable.
View return policy
Description
DLL1 Polyclonal specifically detects DLL1 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
DLL1 | |
Polyclonal | |
Unconjugated | |
P97677 | |
DLL1 | |
Mouse myeloma cell line NS0-derived recombinant rat DLL1 Gln18-Trp537 (predicted) Accession # P97677 | |
100 μg | |
Primary | |
Detects rat and mouse DLL1 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human DLL-1 is observed. | |
Mouse, Rat | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
Delta, delta (Drosophila)-like 1, Delta 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
Sheep | |
Affinity Purified | |
RUO | |
28514 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction