Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat Draxin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6149SP
This item is not returnable.
View return policy
Description
Draxin/C1orf187 Polyclonal specifically detects Draxin/C1orf187 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Draxin/C1orf187 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AGPA3119, C1orf187, chromosome 1 open reading frame 187, dorsal inhibitory axon guidance protein, Dorsal repulsive axon guidance protein, draxin, FLJ34999, MGC117222, Neucrin, UNQ3119 | |
Sheep | |
Affinity Purified | |
RUO | |
374946 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q6PAL1 | |
DRAXIN | |
Mouse myeloma cell line NS0-derived recombinant mouse Draxin Gly26-Ile343 Accession # Q6PAL1 | |
25 μg | |
Primary | |
Detects mouse and rat Draxin in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human Draxin is observed. | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction