Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat FGF basic/FGF2/bFGF Quantikine ELISA Kit

Catalog No. mfb00
Click to view available options
Quantity:
1 x 96-well Plate
This item is not returnable. View return policy
This item is not returnable. View return policy

Mouse/Rat FGF basic/FGF2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse/Rat FGF basic/FGF2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat FGF basic/FGF2 in cell culture supernates (50 ul), tissue lysates (10 ul), serum (50 ul), edta plasma (50 ul).

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 3.68pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum, Tissue Lysates
Specificity Natural and Recombinant FGF basic. This assay cross-reacts 100% with Bovine FGF basic, Recombinant Human FGF basic is detectable.
For Use With (Application) ELISA
Gene ID (Entrez) 14173
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Tissue Lysates (10uL),Serum (50uL),EDTA Plasma (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.