Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat GRIN1/NMDAR1 C1 Splice Variant Antibody, R&D Systems™

Catalog No. PPS080
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS080 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS080 Supplier R&D Systems Supplier No. PPS080
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NMDA R, NR1 Subunit Polyclonal specifically detects NMDA R, NR1 Subunit in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NMDA R, NR1 Subunit
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:1000 - 1:2000 dilution
Gene Alias GluN1, MRD8, NDHMSD, NMDA R, NR1 Subunit, NMDA1, NMDAR1, NMD-R1, NR1
Gene Symbols Grin1
Host Species Rabbit
Immunogen Peptide from the NR1 subunit, C1 Splice Variant of the NMDA Receptor
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2902
Test Specificity Mouse and rat ∼120 kDa NR1 subunit of the NMDA Receptor containing the C1 splice variant
Reconstitution Reconstitute with 50 μL of sterile PBS.
Target Species Mouse, Rat
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.