Learn More
Mouse/Rat HIN-1/SCGB3A1 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | HIN-1/SCGB3A1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL |
| Gene Accession No. | Q920D7 |
| Gene Alias | cytokine high in normal-1, Cytokine HIN-1, High in normal 1, HIN1, HIN-1, HIN1Pneumo secretory protein 2, LU105, PnSP-2, PNSP2, PnSP-2Uteroglobin-related protein 2, SCGB3A1, secretoglobin family 3A member 1, secretoglobin, family 3A, member 1, UGRP2, UGRP2MGC87867 |
| Gene Symbols | Otud4 |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.