Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat ICAM-5 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
$200.50 - $549.50
Specifications
Antigen | ICAM-5 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.2 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
ICAM-5 Polyclonal specifically detects ICAM-5 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ICAM-5 | |
Western Blot 0.2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse, Rat | |
ICAM5, ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, Telencephalin, TLCNTelencephalin, TLNtelencephalin | |
ICAM5 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Q60625 | |
7087 | |
Mouse myeloma cell line NS0-derived recombinant mouse ICAM-5 Leu31-Arg828 (Pro47Arg) Accession # Q60625 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse ICAM-5 in direct ELISAs. Detects mouse and rat ICAM-5 in Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) ICAM-5 and recombinant mouse (rm) ICAM-2 is observed, 5% cross-reactivity with recombinant rat ICAM-1, rhICAM-1, and rhICAM-3 is observed and less than 1% cross-reactivity with rmICAM-1 and rmVCAM-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title