Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat IL-10 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 16 publications
Supplier: R&D Systems AF519
This item is not returnable.
View return policy
Description
IL-10 Polyclonal specifically detects IL-10 in Mouse, Rat samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.Specifications
IL-10 | |
Polyclonal | |
Unconjugated | |
P29456 | |
IL10 | |
E. coli-derived recombinant rat IL-10 Ser19-Asn178 Accession # P29456 | |
100 μg | |
Primary | |
Detects mouse, rat, and cotton rat IL-10 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human IL-10, recombinant guinea pig IL-10, recombinant viral IL-10 and recombinant canine IL-10 is observed. | |
Mouse, Rat | |
IgG |
Western Blot, Neutralization, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.3-1.0 ug/mL, Immunocytochemistry 5-15 ug/mL | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
Goat | |
Affinity Purified | |
RUO | |
3586 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction