Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat IL-5 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF545
This item is not returnable.
View return policy
Description
IL-5 Polyclonal specifically detects IL-5 in Mouse, Rat samples. It is validated for Western Blot.Specifications
IL-5 | |
Polyclonal | |
Biotin | |
Q08125 | |
IL5 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat IL-5 Met20-Val132 Accession # Q08125 | |
50 μg | |
Primary | |
Detects rat and mouse IL-5 in Western blots. In Western blots, less than 2% cross-reactivity with recombinant human IL-5 is observed. | |
Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I | |
Goat | |
Affinity Purified | |
RUO | |
3567 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction