Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Integrin alpha 8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4076
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4076 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4076 Supplier R&D Systems Supplier No. AF4076
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 9 publications

Integrin alpha 8 Polyclonal specifically detects Integrin alpha 8 in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen Integrin alpha 8
Applications Immunocytochemistry, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 μg/mL, Simple Western 10 μg/mL, Immunocytochemistry 5 to 15 μg/mL
Gene Accession No. A28ARA8
Gene Alias integrin alpha-8, integrin, alpha 8, ITGA8
Gene Symbols ITGA8
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 8 Phe38-Phe1007 Accession # A28ARA8
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8516
Test Specificity Detects mouse and rat Integrin alpha 8 in Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human (rh) Integrin alpha 8 is observed, and less than 1% cross-reactivity with rhIntegrin alpha 5, recombinant mouse (rm) Integrin alpha 5, rhIntegrin alpha V and rmIntegrin alpha V is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.