Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat M-Cadherin/Cadherin-15 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB76771
This item is not returnable.
View return policy
Description
M-Cadherin/Cadherin-15 Monoclonal specifically detects M-Cadherin/Cadherin-15 in Mouse, Rat samples. It is validated for Western Blot.Specifications
M-Cadherin/Cadherin-15 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
P33146 | |
CDH15 | |
Mouse myeloma cell line NS0-derived recombinant mouse M-Cadherin/Cadherin-15 Val22-Ala605 Accession # P33146 | |
100 μg | |
Primary | |
Detects mouse M-Cadherin/Cadherin-15 in direct ELISAs and mouse and rat M-Cadherin/Cadherin-15 in Western blots. In direct ELISAs, no cross-reactivity with recombinant human M-Cadherin, recombinant mouse (rm) Cadherin-13, or rmP-Cadherin is observed. | |
Mouse, Rat | |
Purified |
Western Blot | |
800512 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cadherin 15, M-cadherin (myotubule), cadherin 15, type 1, M-cadherin (myotubule), Cadherin-14, Cadherin-15, cadherin-3, CDH14, CDH14cadherin-14, CDH15, CDH3cadherin-15, CDHM, MCAD, M-cadherin, MCadherin, MRD3, Muscle cadherin, muscle-cadherin | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1013 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction