Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat M-Cadherin/Cadherin-15 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7677
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7677 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7677 Supplier R&D Systems Supplier No. AF7677
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

M-Cadherin/Cadherin-15 Polyclonal specifically detects M-Cadherin/Cadherin-15 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen M-Cadherin/Cadherin-15
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P33146
Gene Alias cadherin 15, M-cadherin (myotubule), cadherin 15, type 1, M-cadherin (myotubule), Cadherin-14, Cadherin-15, cadherin-3, CDH14, CDH14cadherin-14, CDH15, CDH3cadherin-15, CDHM, MCAD, M-cadherin, MCadherin, MRD3, Muscle cadherin, muscle-cadherin
Gene Symbols CDH15
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse M-Cadherin/Cadherin-15 Val22-Ala605 (Arg159His) Accession # P33146
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1013
Test Specificity Detects mouse and rat M-Cadherin/Cadherin-15 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human M-Cadherin/Cadherin-15 is observed, and less than 1% cross-reactivity with recombinant mouse P-Cadherin is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.