Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat MIF Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB19781SP
This item is not returnable.
View return policy
Description
MIF Monoclonal specifically detects MIF in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
MIF | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
P34884 | |
MIF | |
E. coli-derived recombinant mouse MIF Met1-Ala115 Accession # P34884 | |
25 μg | |
Primary | |
Detects mouse MIF in ELISA and Western Blot. | |
Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
811409 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4282 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction