Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat MMP-8 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3245
This item is not returnable.
View return policy
Description
MMP-8 Polyclonal specifically detects MMP-8 in Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| MMP-8 | |
| Polyclonal | |
| Unconjugated | |
| O88766 | |
| MMP8 | |
| Mouse myeloma cell line NS0-derived recombinant rat MMP-8 Leu21-Pro466 Accession # O88766 | |
| 100 μg | |
| Primary | |
| Detects rat and mouse MMP-8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human MMP-8 is observed. | |
| Mouse, Rat | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Immunoprecipitation 25 ug/mL | |
| CLG1HNC, Collagenase 2, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4317 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction