Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Nesfatin-1/Nucleobindin-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6895
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6895 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6895 Supplier R&D Systems Supplier No. AF6895
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 2 publications

Nesfatin-1/Nucleobindin-2 Polyclonal specifically detects Nesfatin-1/Nucleobindin-2 in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen Nesfatin-1/Nucleobindin-2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q9JI85
Gene Alias DNA-binding protein NEFA, NEFA, NEFAGastric cancer antigen Zg4, Nesfatin1, NUCB2, nucleobindin 2, nucleobindin-2, nucleobinding 2
Gene Symbols NUCB2
Host Species Sheep
Immunogen E. coli-derived recombinant rat Nesfatin-1/Nucleobindin-2 Pro26-Leu106 Accession # Q9JI85
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4925
Test Specificity Detects rat Nesfatin-1/Nucleobindin-2 in direct ELISAs and mouse and rat Nesfatin-1/Nucleobindin-2 in Western blots. In direct ELISAs, approximately 7% cross-reactivity with recombinant human Nesfatin-1/Nucleobindin-2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.