Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Neuroglycan C/CSPG5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5665SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5665SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5665SP Supplier R&D Systems Supplier No. AF5665SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Neuroglycan C/CSPG5 Polyclonal specifically detects Neuroglycan C/CSPG5 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Neuroglycan C/CSPG5
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH55736
Gene Alias CALEB, chondroitin sulfate proteoglycan 5, chondroitin sulfate proteoglycan 5 (neuroglycan C), CSPG5, MGC44034, Neuroglycan C, NGC, NGCAcidic leucine-rich EGF-like domain-containing brain protein
Gene Symbols CSPG5
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Neuroglycan C/CSPG5 Val31-Gln420 Accession # AAH55736
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10675
Test Specificity Detects mouse and rat Neuroglycan C/CSPG5 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human Neuroglycan C is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.