Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Neuropilin-1 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB566P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB566P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB566P Supplier R&D Systems Supplier No. FAB566P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Neuropilin-1 Polyclonal specifically detects Neuropilin-1 in Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Neuropilin-1
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q9QWJ9
Gene Alias BDCA4, BDCA-4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, Neuropilin1, NP1, NRP, NRP1, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R
Gene Symbols NRP1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-1 Phe22-Ala810 (Lys811Arg), Ser829-Asp854 Accession # Q9QWJ9
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8829
Test Specificity Detects mouse and rat Neuropilin-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 8% cross-reactivity with recombinant human Neuropilin-1 is observed, and less than 1% cross-reactivity with recombinant rat Neuropilin-2 is observed.
Target Species Mouse, Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.