Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat NrCAM Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF8538
This item is not returnable.
View return policy
Description
NrCAM Polyclonal specifically detects NrCAM in Mouse, Rat samples. It is validated for Western Blot.Specifications
NrCAM | |
Polyclonal | |
Western Blot 1:1000 dilution | |
Q810U4 | |
NRCAM | |
Mouse myeloma cell line NS0-derived recombinant mouse NrCAM Leu30-Pro630 Accession # Q810U4 | |
100 μL | |
Primary | |
Detects mouse and rat NrCAM in direct ELISAs and Western blots. In direct ELISAs, less than 25% cross-reactivity with recombinant human NrCAM is observed. | |
12 months from date of receipt, -20 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 degreesC under sterile conditions after opening. | |
IgG |
Western Blot | |
Unconjugated | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. with Sodium Azide | |
bravo, hBravo, KIAA0343Neuronal surface protein Bravo, MGC138845, MGC138846, neuronal cell adhesion molecule, Ng-CAM-related, NgCAM-related cell adhesion molecule, nr-CAM | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
4897 | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction