Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Phospho-DARPP-32 (T34) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2847
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2847 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2847 Supplier R&D Systems Supplier No. AF2847
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DARPP-32 Polyclonal specifically detects DARPP-32 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen DARPP-32 (p thr34)
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Alias DARPP32, PPP1R1B, protein phosphatase 1, regulatory (inhibitor) subunit 1B, regulatory (inhibitor) subunit 1B (dopamine and cAMPregulated phosphoprotein, DARPP-32)
Gene Symbols PPP1R1B
Host Species Rabbit
Immunogen Phosphopeptide containing mouse DARPP-32 T34 site
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84152
Test Specificity Detects mouse and rat DARPP-32 when phosphorylated at T34 in Western blots. There is weak cross-reactivity with phosphorylated PP1 Inhibitor-1, which contains a nearly identical phosphorylation site. Reactivity with human Phospho-DARPP-32 has not been confirmed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.