Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Phospho-Potassium Channel Kv3.1 (S503) Antibody, R&D Systems™

Catalog No. PPS016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS016 Supplier R&D Systems Supplier No. PPS016
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Potassium Channel Kv3.1 Polyclonal specifically detects Potassium Channel Kv3.1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Potassium Channel Kv3.1 (p ser503)
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:1000 dilution
Gene Alias FLJ41162, FLJ42249, FLJ43491, KCNC1, Kv3.1, KV4, MGC129855, NGK2, potassium voltage-gated channel subfamily C member 1, potassium voltage-gated channel, Shaw-related subfamily, member 1, voltage-gated potassium channel protein KV3.1, Voltage-gated potassium channel subunit Kv3.1, Voltage-gated potassium channel subunit Kv4
Gene Symbols KCNC1
Host Species Rabbit
Immunogen Phosphopeptide corresponding to amino acid residues surrounding the phospho-S503 of Potassium Channel Kv3.1
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3746
Test Specificity Rat ∼100k Voltage Gated Potassium Channel KV3.1-subunit protein phosphorylated at S503
Target Species Mouse, Rat
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.