Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat Plexin A3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4075SP
This item is not returnable.
View return policy
Description
Plexin A3 Polyclonal specifically detects Plexin A3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Plexin A3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HSSEXGENE, plexin A3, Plexin-4, plexin-A3, PLXN3, PLXN4, PLXNA3, Semaphorin receptor SEX, SEX, Sex chromosome X transmembrane protein of HGF receptor family 3, SEXPLEXIN-A3,6.3, XAP-6 | |
Goat | |
Affinity Purified | |
RUO | |
55558 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
P70208 | |
PLXNA3 | |
E. coli-derived recombinant mouse Plexin A3 Leu34-Asp150 Accession # P70208 | |
25 μg | |
Primary | |
Detects mouse Plexin A3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Plexin A1, rmPlexin A2, recombinant human (rh) Plexin B1, rhPlexin C1, and rhPlexin D1 is observed. | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction