Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat/Porcine/Equine CD44 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF6127
This item is not returnable.
View return policy
Description
CD44 Polyclonal specifically detects CD44 in Mouse, Rat, Porcine, Equine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready.Specifications
CD44 | |
Polyclonal | |
Unconjugated | |
NP_033981 | |
CD44 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CD44 Gln25-Thr224 Accession # NP_033981 | |
100 μg | |
Primary | |
Detects mouse and rat CD44 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human CD44 is observed. | |
Mouse, Rat, Pig, Equine | |
IgG |
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
CD44, CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCAM, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2, MDU2CD44 antigen (homing function and Indian blood group system), MDU3, MDU3CDW44, MIC4, MIC4MGC10468, MUTCH-I, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
Sheep | |
Affinity Purified | |
RUO | |
960 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction