Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat PRAT4B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5015
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5015 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5015 Supplier R&D Systems Supplier No. AF5015
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PRAT4B/CNPY4 Polyclonal specifically detects PRAT4B/CNPY4 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen PRAT4B/CNPY4
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Gene Accession No. Q8BQ47
Gene Alias canopy 4 homolog (zebrafish), CNPY4, MGC40499PRAT4BPRotein Associated with Tlr4, protein canopy homolog 4
Gene Symbols CNPY4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derive recombinant mouse PRAT4B Thr30-Leu245 Accession # Q8BQ47
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245812
Test Specificity Detects mouse and rat PRAT4B in Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human PRAT4B and recombinant mouse PRAT4A is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.